rac-Fmoc-Trifluoromethylalanine

Leverandør: Apollo Scientific

PC27870-100MG PC27870-1G PC27870-250MG
APOSPC27870-100MGEA 8775 NOK
APOSPC27870-100MG APOSPC27870-1G APOSPC27870-250MG
rac-Fmoc-Trifluoromethylalanine
rac-Fmoc-Trifluoromethylalanine
Formel: C₁₈H₁₄F₃NO₄
MW: 365.303 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28898379
CAS-nummer: 1219349-78-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR