3-Chloro-2-(1,2-oxazol-4-yl)-5-(trifluoromethyl)pyridine 95%

Leverandør: Apollo Scientific

PC27944-1G
APOSPC27944-1GEA 19275 NOK
APOSPC27944-1G
3-Chloro-2-(1,2-oxazol-4-yl)-5-(trifluoromethyl)pyridine 95%
3-Chloro-2-(1,2-oxazol-4-yl)-5-(trifluoromethyl)pyridine
Formel: C₉H₄ClF₃N₂O
MW: 248,589 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00231890
CAS-nummer: 246022-17-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR