1,1,1-Trifluoro-3-(4-phenylpiperazin-1-yl)propan-2-ol, tech

Leverandør: Apollo Scientific

PC28122-1G
APOSPC28122-1GEA 19275 NOK
APOSPC28122-1G
1,1,1-Trifluoro-3-(4-phenylpiperazin-1-yl)propan-2-ol, tech
1,1,1-Trifluoro-3-(4-phenylpiperazin-1-yl)propan-2-ol
Formel: C₁₃H₁₇F₃N₂O
MW: 274,282 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02083036
CAS-nummer: 672952-04-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR