(2Z)-2-[(4-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Leverandør: Apollo Scientific

PC28142-1G
APOSPC28142-1GEA 19275 NOK
APOSPC28142-1G
(2Z)-2-[(4-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2Z)-2-[(4-Fluorophenyl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₄H₁₄FNO
MW: 231,265 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00440969
CAS-nummer: 209863-77-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR