2-(Trifluoromethyl)benzenesulphonamide

Leverandør: Apollo Scientific
Warning

Synonymer: α,α,α-Trifluoro-o-toluenesulphonamide

PC2977-1G PC2977-25G PC2977-5G
APOSPC2977-1GEA 270 NOK
APOSPC2977-1G APOSPC2977-25G APOSPC2977-5G
2-(Trifluoromethyl)benzenesulphonamide
2-(Trifluoromethyl)benzenesulphonamide
Formel: C₇H₆F₃NO₂S
MW: 225.19
Smeltepunkt: 180…184 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00042420
CAS-nummer: 1869-24-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR