4-(Trifluoromethyl)benzenesulphonamide

Leverandør: Apollo Scientific
Warning

Synonymer: α,α,α-Trifluoro-p-toluenesulphonamide

PC2981-25G PC2981-5G
APOSPC2981-25GEA 805 NOK
APOSPC2981-25G APOSPC2981-5G
4-(Trifluoromethyl)benzenesulphonamide
4-(Trifluoromethyl)benzenesulphonamide
Formel: C₇H₆F₃NO₂S
MW: 225.19
Smeltepunkt: 181 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00159251
CAS-nummer: 830-43-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR