Ethyl 1-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-4-carboxylate

Leverandør: Apollo Scientific

PC300881-1G
APOSPC300881-1GEA 4175 NOK
APOSPC300881-1G
Ethyl 1-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-4-carboxylate
Ethyl 1-(3,3,3-trifluoro-2-hydroxypropyl)piperidine-4-carboxylate
Formel: C₁₁H₁₈F₃NO₃
MW: 269,261 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02186262
CAS-nummer: 478258-74-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR