Ethyl 2-{[4-(trifluoromethoxy)phenyl]hydrazono}propanoate

Leverandør: Apollo Scientific

PC300957-1G
APOSPC300957-1GEA 2800 NOK
APOSPC300957-1G
Ethyl 2-{[4-(trifluoromethoxy)phenyl]hydrazono}propanoate
Ethyl 2-{[4-(trifluoromethoxy)phenyl]hydrazono}propanoate
Formel: C₁₂H₁₃F₃N₂O₃
MW: 290,238 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00219835
CAS-nummer: 883006-93-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR