Methyl 2-(3-fluorophenoxy)propanoate

Leverandør: Apollo Scientific

PC301017-1G
APOSPC301017-1GEA 3825 NOK
APOSPC301017-1G
Methyl 2-(3-fluorophenoxy)propanoate
Methyl 2-(3-fluorophenoxy)propanoate
Formel: C₁₀H₁₁FO₃
MW: 198,191 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD16042155
CAS-nummer: 1247636-15-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR