2,3-Difluoro-p-toluidine

Leverandør: Apollo Scientific
Warning

PC302637-5G PC302637-1G
APOSPC302637-5GEA 1775 NOK
APOSPC302637-5G APOSPC302637-1G
2,3-Difluoro-p-toluidine
2,3-Difluoro-p-toluidine
Formel: C₇H₇F₂N
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06660321
CAS-nummer: 886503-79-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR