5-(Trifluoromethoxy)-m-toluidine 98%

Leverandør: Apollo Scientific
Warning

PC302962-1G PC302962-5G PC302962-10G
APOSPC302962-1GEA 2850 NOK
APOSPC302962-1G APOSPC302962-5G APOSPC302962-10G
5-(Trifluoromethoxy)-m-toluidine 98%
5-(Trifluoromethoxy)-m-toluidine
Formel: C₈H₈F₃NO
Kokepunkt: 73 °C/1mmHg
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09025380
CAS-nummer: 86256-63-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR