4-Amino-2-fluorophenylboronic acid pinacol ester

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Amino-2-fluorobenzeneboronic acid pinacol ester, 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

PC3221-5G
APOSPC3221-5GEA 1200 NOK
APOSPC3221-5G
4-Amino-2-fluorophenylboronic acid pinacol ester
4-Amino-2-fluorophenylboronic acid pinacol ester
Formel: C₁₂H₁₇BFNO₂
MW: 237.08 g/mol
Smeltepunkt: 163…166 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD09951877
CAS-nummer: 819057-45-9

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR