(2E)-3-{[3-(Trifluoromethyl)phenyl]carbamoyl}prop-2-enoic acid

Leverandør: Apollo Scientific

PC32260-1G
APOSPC32260-1GEA 1450 NOK
APOSPC32260-1G
(2E)-3-{[3-(Trifluoromethyl)phenyl]carbamoyl}prop-2-enoic acid
(2E)-3-{[3-(Trifluoromethyl)phenyl]carbamoyl}prop-2-enoic acid
Formel: C₁₁H₈F₃NO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00029807
CAS-nummer: 53616-18-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR