3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole

Leverandør: Apollo Scientific

PC3652-1G
APOSPC3652-1GEA 2175 NOK
APOSPC3652-1G
3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole
3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole
Formel: C₁₄H₈BrFN₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09878375
CAS-nummer: 1000339-25-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR