3-Fluoro-p-toluidine 98%

Leverandør: Apollo Scientific
Danger

Synonymer: 3-Fluoro-4-methylaniline, 4-Amino-2-fluorotoluene

PC3818-100G PC3818-25G
APOSPC3818-100GEA 965 NOK
APOSPC3818-100G APOSPC3818-25G
3-Fluoro-p-toluidine 98%
3-Fluoro-p-toluidine
Formel: C₇H₈FN
Kokepunkt: 93 °C/12mmHg
Smeltepunkt: 30…35 °C
Tetthet: 1,093 g/cm³ (25 °C)
Flash punkt: 88 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007762
CAS-nummer: 452-77-7
EINECS: 207-212-4
FN: 2811
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR