Methyl 6-fluoro-4-(methylamino)pyridine-3-carboxylate

Leverandør: Apollo Scientific
Warning

PC400036-1G
APOSPC400036-1GEA 5875 NOK
APOSPC400036-1G
Methyl 6-fluoro-4-(methylamino)pyridine-3-carboxylate
Methyl 6-fluoro-4-(methylamino)pyridine-3-carboxylate
Formel: C₈H₉FN₂O₂
MW: 184,168 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD31536751
CAS-nummer: 2187435-19-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR