4-Bromo-5-(difluoromethyl)-1H-pyrazole

Leverandør: Apollo Scientific
Warning

PC400542-1G
APOSPC400542-1GEA 8275 NOK
APOSPC400542-1G
4-Bromo-5-(difluoromethyl)-1H-pyrazole
4-Bromo-5-(difluoromethyl)-1H-pyrazole
Formel: C₄H₃BrF₂N₂
MW: 196,981 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD23704677
CAS-nummer: 1451392-65-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR