(2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-enal

Leverandør: Apollo Scientific

PC401049-1G
APOSPC401049-1GEA 4300 NOK
APOSPC401049-1G
(2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-enal
(2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-enal
Formel: C₁₁H₁₀ClF₃N₂O
MW: 278,658 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00231994
CAS-nummer: 339010-44-5

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR