3-(Aminomethyl)-6-fluoro-1H-indole 96%

Leverandør: Apollo Scientific
Warning

Synonymer: 1-(6-Fluoro-1H-indol-3-yl)methylamine

PC402016-1G
APOSPC402016-1GEA 12100 NOK
APOSPC402016-1G
3-(Aminomethyl)-6-fluoro-1H-indole 96%
3-(Aminomethyl)-6-fluoro-1H-indole
Formel: C₉H₉FN₂
MW: 164,18 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06657116
CAS-nummer: 887582-19-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR