(DL)-4-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid

Leverandør: Apollo Scientific

PC405603-100MG
APOSPC405603-100MGEA 16000 NOK
APOSPC405603-100MG
(DL)-4-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid
(DL)-4-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid
Formel: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 1404192-14-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR