1-Fluoro-3-isocyanato-2-methylbenzene 95%

Leverandør: Apollo Scientific

PC408064-5G
APOSPC408064-5GEA 1875 NOK
APOSPC408064-5G
1-Fluoro-3-isocyanato-2-methylbenzene 95%
1-Fluoro-3-isocyanato-2-methylbenzene

May form small amount of sediment

Formel: C₈H₆FNO
MW: 151.138 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD07779361
CAS-nummer: 60221-81-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR