2-(4-Chloromethyl-piperidin-1-yl)-5-fluoro-pyrimidine

Leverandør: Apollo Scientific

Synonymer: 2-(4-(Chloromethyl)piperidin-1-yl)-5-fluoropyrimidine

PC408355-500MG
APOSPC408355-500MGEA 7625 NOK
APOSPC408355-500MG
2-(4-Chloromethyl-piperidin-1-yl)-5-fluoro-pyrimidine
2-(4-Chloromethyl-piperidin-1-yl)-5-fluoro-pyrimidine
Formel: C₁₀H₁₃ClFN₃
MW: 229,682 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD21098890
CAS-nummer: 1353966-24-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR