2-[(4-Fluoro-benzenesulphonylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester

Leverandør: Apollo Scientific

Synonymer: tert-Butyl 2-((4-fluorophenylsulphonamido)methyl)piperidine-1-carboxylate

PC408377-500MG
APOSPC408377-500MGEA 7625 NOK
APOSPC408377-500MG
2-[(4-Fluoro-benzenesulphonylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
2-[(4-Fluoro-benzenesulphonylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₅FN₂O₄S
MW: 372,455 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD21098973
CAS-nummer: 1353980-93-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR