4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide

Leverandør: Apollo Scientific

PC410291-1G
APOSPC410291-1GEA 2875 NOK
APOSPC410291-1G
4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
Formel: C₆H₆ClF₃N₄O
MW: 242,586 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03419819
CAS-nummer: 1001519-39-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR