1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-3-amine

Leverandør: Apollo Scientific

PC410296-1G
APOSPC410296-1GEA 4750 NOK
APOSPC410296-1G
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-3-amine
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-3-amine
Formel: C₁₀H₉ClFN₃
MW: 225,65 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02254006
CAS-nummer: 1001757-50-1

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR