4-Chloro-1-(3-fluorobenzyl)-1H-pyrazol-3-amine

Leverandør: Apollo Scientific

PC410299-1G
APOSPC410299-1GEA 6350 NOK
APOSPC410299-1G
4-Chloro-1-(3-fluorobenzyl)-1H-pyrazol-3-amine
4-Chloro-1-(3-fluorobenzyl)-1H-pyrazol-3-amine
Formel: C₁₀H₉ClFN₃
MW: 225,65 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02254024
CAS-nummer: 1001757-55-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR