2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

Leverandør: Apollo Scientific

PC410408-1G
APOSPC410408-1GEA 7975 NOK
APOSPC410408-1G
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Formel: C₉H₁₁F₃N₂O₂
MW: 236,191 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD04967809
CAS-nummer: 1006348-57-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR