2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile

Leverandør: Apollo Scientific

PC410422-1G
APOSPC410422-1GEA 4750 NOK
APOSPC410422-1G
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
2-Methyl-3-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]propanenitrile
Formel: C₉H₁₀F₃N₃
MW: 217,191 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD08556255
CAS-nummer: 1006352-92-6

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR