cis-3-Fluorocyclobutane-1-carboxylic acid

Leverandør: Apollo Scientific

Synonymer: cis-3-Fluorocyclobutanecarboxylic acid

PC411271-1G
APOSPC411271-1GEA 34700 NOK
APOSPC411271-1G
cis-3-Fluorocyclobutane-1-carboxylic acid
cis-3-Fluorocyclobutane-1-carboxylic acid
Formel: C₅H₇FO₂
MW: 118.106 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD16661179
CAS-nummer: 123812-78-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR