3-Cyano-4-fluorophenylboronic acid pinacol ester 95+%

Leverandør: Apollo Scientific

Synonymer: 3-Cyano-4-fluorobenzeneboronic acid pinacol ester, 2-(3-Cyano-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Cyano-4-fluorophenylboronic acid Pinacol ester, 3-Cyano-4-fluorophenylboronic acid, pinacol ester, 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

PC412081-25G
APOSPC412081-25GEA 2300 NOK
APOSPC412081-25G
3-Cyano-4-fluorophenylboronic acid pinacol ester 95+%
3-Cyano-4-fluorophenylboronic acid pinacol ester
Formel: C₁₃H₁₅BFNO₂
MW: 247.08 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD06795681
CAS-nummer: 775351-57-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR