8-Boc-2-fluoro-8-aza-bicyclo[3.2.1]octan-3-one

Leverandør: Apollo Scientific
Danger

Synonymer: 2-Methyl-2-propanyl 2-fluoro-3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate

PC430197-1G
APOSPC430197-1GEA 4475 NOK
APOSPC430197-1G
8-Boc-2-fluoro-8-aza-bicyclo[3.2.1]octan-3-one
8-Boc-2-fluoro-8-aza-bicyclo[3.2.1]octan-3-one
Formel: C₁₂H₁₈FNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22580379
CAS-nummer: 1404196-37-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR