Ethyl 3-(benzyl(2-ethoxy-2-oxoethyl)amino)-2,2-difluoropropanoate

Leverandør: Apollo Scientific

PC430238-1G
APOSPC430238-1GEA 10650 NOK
APOSPC430238-1G
Ethyl 3-(benzyl(2-ethoxy-2-oxoethyl)amino)-2,2-difluoropropanoate
Ethyl 3-(benzyl(2-ethoxy-2-oxoethyl)amino)-2,2-difluoropropanoate
Formel: C₁₆H₂₁F₂NO₄
MW: 329,339 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28016237
CAS-nummer: 1823791-33-8

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR