2-(8-Aza-bicyclo[3.2.1]octan-3-yl)acetic acid 2,2,2-trifluoroacetate

Leverandør: Apollo Scientific

PC430315-1G
APOSPC430315-1GEA 10650 NOK
APOSPC430315-1G
2-(8-Aza-bicyclo[3.2.1]octan-3-yl)acetic acid 2,2,2-trifluoroacetate
2-(8-Aza-bicyclo[3.2.1]octan-3-yl)acetic acid 2,2,2-trifluoroacetate
Formel: C₉H₁₅NO₂·CF₃CO₂H
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD27978598
CAS-nummer: 1980044-86-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR