4-Acetamido-3-fluorobenzene-1-sulphonyl chloride

Leverandør: Apollo Scientific

PC430542-100MG
APOSPC430542-100MGEA 4150 NOK
APOSPC430542-100MG
4-Acetamido-3-fluorobenzene-1-sulphonyl chloride
4-Acetamido-3-fluorobenzene-1-sulphonyl chloride
Formel: C₈H₇ClFNO₃S
MW: 251,662 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00052328
CAS-nummer: 349-71-3

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR