N-Allyl-2-(3-(trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)acetamide

Leverandør: Apollo Scientific

PC430601-500MG
APOSPC430601-500MGEA 9300 NOK
APOSPC430601-500MG
N-Allyl-2-(3-(trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)acetamide
N-Allyl-2-(3-(trifluoromethyl)-4,5,6,7-tetrahydroindazol-1-yl)acetamide
Formel: C₁₃H₁₆F₃N₃O
MW: 287,281 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD18157704
CAS-nummer: 1258652-68-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR