p-Nitrophenyl 2-(m-trifluoromethylphenyl)-7-aza-spiro[3.5]nonane-7-carboxylate

Leverandør: Apollo Scientific

PC430605-250MG
APOSPC430605-250MGEA 11600 NOK
APOSPC430605-250MG
p-Nitrophenyl 2-(m-trifluoromethylphenyl)-7-aza-spiro[3.5]nonane-7-carboxylate
p-Nitrophenyl 2-(m-trifluoromethylphenyl)-7-aza-spiro[3.5]nonane-7-carboxylate
Formel: C₂₂H₂₁F₃N₂O₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD20926407
CAS-nummer: 1225276-30-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR