2,2,2-Trifluoro-2',4',6'-trimethylacetophenone 97%

Leverandør: Apollo Scientific
Warning

Synonymer: 2,2,2-Trifluoro-1-mesitylethanone

PC4354-1G
APOSPC4354-1GEA 6175 NOK
APOSPC4354-1G
2,2,2-Trifluoro-2',4',6'-trimethylacetophenone 97%
2,2,2-Trifluoro-2',4',6'-trimethylacetophenone
Formel: C₁₁H₁₁F₃O
MW: 216,19 g/mol
Kokepunkt: 78…80 °C/20mmHg
Tetthet: 1,136 g/cm³ (25 °C)
Flash punkt: 76 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00151856
CAS-nummer: 313-56-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR