2-Fluoro-4-(trifluoromethyl)benzylamine 97%

Leverandør: Apollo Scientific
Danger

Synonymer: (2-Fluoro-4-(trifluoromethyl)phenyl)methanamine

PC4374U-500MG PC4374U-250MG
APOSPC4374U-500MGEA 305 NOK
APOSPC4374U-500MG APOSPC4374U-250MG
2-Fluoro-4-(trifluoromethyl)benzylamine 97%
2-Fluoro-4-(trifluoromethyl)benzylamine
Formel: C₈H₇F₄N
MW: 193.14 g/mol
Kokepunkt: 186
Smeltepunkt: 46
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070807
CAS-nummer: 239087-05-9
FN: 2735
ADR: 8 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR