2-Fluoro-6-(trifluoromethyl)benzylamine 95%

Leverandør: Apollo Scientific
Danger

Synonymer: 2-(Aminomethyl)-3-fluorobenzotrifluoride, [2-Fluoro-6-(trifluoromethyl)phenyl]methylamine

PC4374W-1G PC4374W-5G PC4374W-25G
APOSPC4374W-1GEA 270 NOK
APOSPC4374W-1G APOSPC4374W-5G APOSPC4374W-25G
2-Fluoro-6-(trifluoromethyl)benzylamine 95%
2-Fluoro-6-(trifluoromethyl)benzylamine
Formel: C₈H₇F₄N
MW: 193.14 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00061165
CAS-nummer: 239087-06-0
FN: 2735
ADR: 8 III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR