4-Fluoro-2-(trifluoromethyl)benzylamine 97%

Leverandør: Apollo Scientific
Danger

Synonymer: (4-Fluoro-2-(trifluoromethyl)phenyl)methanamine

PC4374Y-25G PC4374Y-5G PC4374Y-100G
APOSPC4374Y-25GEA 1700 NOK
APOSPC4374Y-25G APOSPC4374Y-5G APOSPC4374Y-100G
4-Fluoro-2-(trifluoromethyl)benzylamine 97%
4-Fluoro-2-(trifluoromethyl)benzylamine
Formel: C₈H₇F₄N
MW: 193.14 g/mol
Kokepunkt: 185
Smeltepunkt: 46
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00061164
CAS-nummer: 202522-22-3
FN: 2735
ADR: 8 II
New Product

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR