4-Fluoro-3-(trifluoromethyl)benzylamine 97%

Leverandør: Apollo Scientific
Danger

Synonymer: (4-Fluoro-3-(trifluoromethyl)phenyl)methanamine

PC4374Z-25G PC4374Z-5G
APOSPC4374Z-25GEA 4175 NOK
APOSPC4374Z-25G APOSPC4374Z-5G
4-Fluoro-3-(trifluoromethyl)benzylamine 97%
4-Fluoro-3-(trifluoromethyl)benzylamine
Formel: C₈H₇F₄N
MW: 193.14 g/mol
Kokepunkt: 101 °C/3mmHg
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070814
CAS-nummer: 67515-74-6
FN: 2735
ADR: 8 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR