Galdenr® HT270

Leverandør: Apollo Scientific
Warning

Synonymer: Perfluoropolyether YR-1800

PC4387K-1KG
APOSPC4387K-1KGEA 7975 NOK
APOSPC4387K-1KG
Galdenr® HT270
1-Propene, 1,1,2,3,3,3-hexafluoro-, oxidized, polymerized
Kokepunkt: 270 °C
Smeltepunkt: –66 °C
Tetthet: 1,85 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00748116
CAS-nummer: 69991-67-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR