6-Fluoro-m-toluidine

Leverandør: Apollo Scientific
Warning

Synonymer: 3-Amino-4-fluorotoluene, 6-Fluoro-m-methylaniline, 2-Fluoro-5-methylaniline

PC4433-10G PC4433-250G PC4433-50G
APOSPC4433-10GEA 270 NOK
APOSPC4433-10G APOSPC4433-250G APOSPC4433-50G
6-Fluoro-m-toluidine
6-Fluoro-m-toluidine
Formel: C₇H₈FN
Kokepunkt: 80…86 °C/11mmHg
Tetthet: 1,109 g/cm³ (25 °C)
Flash punkt: 81 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007654
CAS-nummer: 452-84-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR