(1S)-1-(3,5-Difluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate

Leverandør: Apollo Scientific

PC450029-250MG
APOSPC450029-250MGEA 2800 NOK
APOSPC450029-250MG
(1S)-1-(3,5-Difluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
(1S)-1-(3,5-Difluorophenyl)-N-[(1S)-1-phenylethyl]ethanaminium 2-carboxybenzoate
Formel: C₁₆H₁₈F₂N·C₈H₅O₄
MW: 427,441 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD28397825
CAS-nummer: 444643-21-4

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR