2,2-Difluoro-2-(2-methylphenyl)ethan-1-ol

Leverandør: Apollo Scientific

PC450038-100MG
APOSPC450038-100MGEA 3500 NOK
APOSPC450038-100MG
2,2-Difluoro-2-(2-methylphenyl)ethan-1-ol
2,2-Difluoro-2-(2-methylphenyl)ethan-1-ol
Formel: C₉H₁₀F₂O
MW: 172.172 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD22124009
CAS-nummer: 1365808-64-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR