Tris(3-fluoropropyl)phosphate

Leverandør: Apollo Scientific
Warning

PC450091-5G
APOSPC450091-5GEA 11825 NOK
APOSPC450091-5G
Tris(3-fluoropropyl)phosphate
Tris(3-fluoropropyl)phosphate
Formel: C₉H₁₈F₃O₄P
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD20489415
CAS-nummer: 1346521-40-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR