1,1,1,2,3,3-Hexafluoropropyl-2,2-difluoroethyl ether

Leverandør: Apollo Scientific

PC450207-1G
APOSPC450207-1GEA 5300 NOK
APOSPC450207-1G
1,1,1,2,3,3-Hexafluoropropyl-2,2-difluoroethyl ether
1,1,1,2,3,3-Hexafluoropropyl-2,2-difluoroethyl ether
Formel: C₅H₄F₈O
MW: 232,072 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD19442560
CAS-nummer: 1309602-06-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR