Sodium 2,2-difluoropropionate

Leverandør: Apollo Scientific

PC450283-5G
APOSPC450283-5GEA 735 NOK
APOSPC450283-5G
Sodium 2,2-difluoropropionate
Sodium 2,2-difluoropropionate
Formel: C₃H₃F₂O₂·Na
MW: 132,041 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD20489367
CAS-nummer: 1281943-34-7

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR