4-Amino-3',5'-bis(trifluoromethyl)diphenyl ether 95+%

Leverandør: Apollo Scientific
Warning

Synonymer: 4-[3,5-Bis(trifluoromethyl)phenoxy]aniline

PC4722-1G
APOSPC4722-1GEA 1075 NOK
APOSPC4722-1G
4-Amino-3',5'-bis(trifluoromethyl)diphenyl ether 95+%
4-Amino-3',5'-bis(trifluoromethyl)diphenyl ether
Formel: C₁₄H₉F₆NO
Smeltepunkt: 64…66 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00662561
CAS-nummer: 57688-35-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR