4-Fluorobenzoyl chloride, Technical Grade

Leverandør: Apollo Scientific
Danger

Synonymer: p-Fluorobenzoyl chloride

PC48430-100G
APOSPC48430-100GEA 270 NOK
APOSPC48430-100G
4-Fluorobenzoyl chloride, Technical Grade
4-Fluorobenzoyl chloride
Formel: C₇H₄ClFO
MW: 158.56 g/mol
Kokepunkt: 82 °C/20mmHg
Smeltepunkt: 44540 °C
Tetthet: 1.326 g/cm³ (25 °C)
Flash punkt: 82 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000684
CAS-nummer: 403-43-0
EINECS: 206-961-4
FN: 3265
ADR: 8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR